Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF39414146

Tantalum

tube, 200mm, outside diameter 8.7mm, inside diameter 8.34mm, wall thickness 0.18mm, as drawn, 99.9%

Synonym(s):

Tantalum, TA007501, Ta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

<0.01 mmHg ( 537.2 °C)

assay

99.9%

form

tubes

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 394-141-46

resistivity

13.5 μΩ-cm, 20°C

L × wall thickness

200 mm × 0.18 mm

O.D. × I.D.

8.7 mm × 8.34 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
R M Rabinovich
Vestnik rentgenologii i radiologii, (1)(1), 77-79 (1981-01-01)
Clinical use of porous tantalum in complex primary total knee arthroplasty.
Neil P Sheth et al.
American journal of orthopedics (Belle Mead, N.J.), 38(10), 526-530 (2009-12-17)
Metallosis and elevated serum levels of tantalum following failed revision hip arthroplasty--a case report.
George C Babis et al.
Acta orthopaedica, 85(6), 677-680 (2014-08-21)
J Black
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
Robert Cohen
American journal of orthopedics (Belle Mead, N.J.), 31(4), 216-217 (2002-05-15)
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service