Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF35804257

Inconel® alloy 600 - heat resisting alloy, Ni72Cr16Fe8

foil, 150 x 150mm, 5.0mm thickness

Synonym(s):

NI020575

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23
Product GF35804257 is not currently sold in your country. Contact Technical Service

form

foil

manufacturer/tradename

Goodfellow 358-042-57

size × thickness

150 mm × 150 mm × 5.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

INCONEL is a registered trademark of Special Metals Corporation
Inconel is a registered trademark of Special Metals Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service