Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF33974466

Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti

foil, 10mm disks, 0.025mm thickness, half hard

Synonym(s):

FE210247

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

form

foil (disks - half hard)

manufacturer/tradename

Goodfellow 339-744-66

size × thickness

10 mm × 0.025 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service