Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF33428831

Cobalt

microfoil, disks, 10mm, thinness 0.1μm, specific density 94.6μg/cm2, permanent mylar 3.5μm support, 99.9%

Synonym(s):

Cobalt, CO004500

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.90%

form

foil

manufacturer/tradename

Goodfellow 334-288-31

resistivity

6.24 μΩ-cm, 20°C

diam. × thickness

10 mm × 0.1 μm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth M Unice et al.
Chemico-biological interactions, 216, 53-74 (2014-04-15)
An updated biokinetic model for human exposures to cobalt (Co) was developed based on a comprehensive set of human pharmacokinetics data collected from five male and five female volunteers who ingested ∼1 mg Co/day of a Co supplement for 3
K Praveena et al.
Journal of nanoscience and nanotechnology, 14(6), 4371-4376 (2014-04-18)
The Cobalt ferrite (CoFe2O4) powders were synthesized by Co-precipitation method. The as prepared ferrite powders were incorporated into a polyaniline matrix at various volumetric ratios. The as prepared composites of ferrite and polyaniline powders were characterized using X-ray diffraction (XRD)
S Dianat et al.
Chemico-biological interactions, 215, 25-32 (2014-03-13)
The parent and nanosized starch, and lipid encapsulated K6[SiW11O39Co(H2O)]·nH2O (abbreviated as SEP, LEP and SiW11Co, respectively), as potent antitumor candidates, were synthesized and characterized by FT-IR spectroscopy, ICP, TG analysis, SEM and TEM images. The results show that the SiW11Co
Aya Watanabe et al.
Clinical science (London, England : 1979), 126(9), 671-678 (2013-11-22)
In the present study we sought to determine the effect of CoCl2, an inhibitor of PHD (prolyl hydroxylase domain protein), on the development of AAA (abdominal aortic aneurysm). AAA was induced in C57BL/6 mice by periaortic application of CaCl2 (AAA
Dennis J Paustenbach et al.
Critical reviews in toxicology, 43(4), 316-362 (2013-05-10)
Cobalt (Co) is an essential element with ubiquitous dietary exposure and possible incremental exposure due to dietary supplements, occupation and medical devices. Adverse health effects, such as cardiomyopathy and vision or hearing impairment, were reported at peak blood Co concentrations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service