GF31858454
Samarium
foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Synonym(s):
Samarium, SM000230
About This Item
assay
99%
form
foil
manufacturer/tradename
Goodfellow 318-584-54
resistivity
91.4 μΩ-cm, 0°C
size × thickness
50x50 mm × 0.25 mm
bp
1794 °C (lit.)
mp
1074 °C (lit.)
density
7.47 g/mL at 25 °C (lit.)
SMILES string
[Sm]
InChI
1S/Sm
InChI key
KZUNJOHGWZRPMI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Water-react 3
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service