Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF29753411

Carbon

microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, 99.997%

Synonym(s):

Activated charcoal, C 004960, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

assay

99.997%

form

foil (microleaf)

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 297-534-11

resistivity

1375 μΩ-cm, 20°C (graphite)

size × thickness

25 × 70 mm × 0.50 μm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer treatment: Sharp shooters.
Vivien Marx
Nature, 508(7494), 133-138 (2014-04-04)
Anna Iliadi et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 145(4), 425-433 (2014-04-08)
The aim of this study was to assess the effect of intraoral aging on the setting status of a resin composite and a glass ionomer adhesive, relative to control specimens stored in water. Metallic brackets were bonded with resin composite
B Mukherjee et al.
Journal of environmental biology, 35(3), 577-588 (2014-05-13)
Modelling system dynamics in a hyper-eutrophic lake is quite complex especially with a constant influx of detergents and sewage material which continually changes the state variables and interferes with the assessment of the chemical rhythm occurring in polluted conditions as
Kimberly T Barrett et al.
Nature, 509(7498), 71-75 (2014-04-22)
One of the most well-recognized stereogenic elements in a chiral molecule is an sp(3)-hybridized carbon atom that is connected to four different substituents. Axes of chirality can also exist about bonds with hindered barriers of rotation; molecules containing such axes
Deivasigamani Ranjith Kumar et al.
Journal of nanoscience and nanotechnology, 14(7), 5059-5069 (2014-04-25)
Iron oxide (Fe3O4) nanoparticles are prepared by nucleation method using oleylamine (OLA) as the capping agent. The OLA capped Fe3O4 nanoparticles (OLA-Fe3O4) are characterized by Fourier Transform Infrared (FT-IR) spectroscopy, X-ray Diffraction (XRD) and High Resolution Transmission Electron Microscopic (HRTEM)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service