Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF29496426

Zirconium

foil, 25x25mm, thickness 0.25mm, annealed, 99.2%

Synonym(s):

Zirconium, ZR000300

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

assay

99.20%

form

foil

manufacturer/tradename

Goodfellow 294-964-26

resistivity

40 μΩ-cm, 20°C

size × thickness

25x25 mm × 0.25 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yin Zhang et al.
Current radiopharmaceuticals, 4(2), 131-139 (2011-12-24)
Positron emission tomography (PET) imaging with radiolabeled monoclonal antibodies has always been a dynamic area in molecular imaging. With decay half-life (3.3 d) well matched to the circulation half-lives of antibodies (usually on the order of days), (89)Zr has been
Joseph R Osborne et al.
The Journal of urology, 191(5), 1439-1445 (2013-10-19)
In this pilot study we explored the feasibility of (89)Zr labeled J591 monoclonal antibody positron emission tomography of localized prostate cancer. Before scheduled radical prostatectomy 11 patients were injected intravenously with (89)Zr-J591, followed 6 days later by whole body positron
David B N Lee et al.
ASAIO journal (American Society for Artificial Internal Organs : 1992), 56(6), 550-556 (2011-01-20)
Recent years have witnessed a rapid increase in the use of zirconium (Zr)-containing compounds in artificial internal organs. Examples include dental implants and other restorative practices, total knee and hip replacement, and middle-ear ossicular chain reconstruction. In nephrological practice, Zr-containing
Thomas J Heyse et al.
Expert review of medical devices, 9(4), 409-421 (2012-08-22)
Polyethylene wear is a key contributor to long-term failure in total knee arthroplasty. Oxidized zirconium alloy was created as a bearing surface for total joint arthroplasty in an attempt to address this concern. Oxidized zirconium has shown considerable improvements over
D B Zhu et al.
Journal of nanoscience and nanotechnology, 14(5), 3703-3706 (2014-04-17)
Functionalized ceramic dental crown was successfully fabricated through selective slurry extrusion (SSE) based technique of solid freeform fabrication (also known as rapid prototyping). After sintering, the decomposed tourmaline powders were embedded in ZrO2 matrix. The far infrared emission properties of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service