Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF28671059

T1 - thermocouple alloy, Ni90Cr10

insulated wire, 5m length, 0.05mm conductor diameter, 0.002mm insulation thickness, polyester insulation

Synonym(s):

NI095820

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300

description

polyester insulation
thermocouple wire

form

wire (insulated)

manufacturer/tradename

Goodfellow 286-710-59

insulation thickness

0.002 mm

wire L × diam.

5 m × 0.05 mm , conductor diameter

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Application

may be used as one component of a thermocouple

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service