Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF28024545

Silver

insulated wire, conductor diameter 0.04 mm, length 5 m, insulation thickness 0.007 mm

Synonym(s):

AG005820

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$712.00

$712.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$712.00

About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

$712.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

form

wire

manufacturer/tradename

Goodfellow 280-245-45

resistivity

1.59 μΩ-cm, 20°C

L

5 m

diam.

0.04 mm

insulation thickness

0.007 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service