GF22546649
Tantalum
foil, 6mm disks, thickness 0.006mm, 99.9%
Synonym(s):
Tantalum, TA000260, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.01 mmHg ( 537.2 °C)
assay
99.90%
form
foil
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 225-466-49
resistivity
13.5 μΩ-cm, 20°C
diam. × thickness
6 mm × 0.006 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tantalum foil pegs.
E MAXWELL
American journal of ophthalmology, 34(3), 444-444 (1951-03-01)
Radmila Panajotovic et al.
Radiation research, 165(4), 452-459 (2006-04-04)
We determined effective cross sections for production of single-strand breaks (SSBs) in plasmid DNA [pGEM 3Zf(-)] by electrons of 10 eV and energies between 0.1 and 4.7 eV. After purification and lyophilization on a chemically clean tantalum foil, dry plasmid
T Y Hung et al.
Applied optics, 31(22), 4397-4404 (1992-08-01)
A simple four-element fused-silica lens is presented that has a focal length of 31.2 mm and a relative aperture of f/1 for use as a focusing lens for deep UV laser processing. The curvature of the lens is designed with
J G Sen Gupta
Talanta, 31(12), 1053-1056 (1984-12-01)
A 40-fold increase in sensitivity obtained by using a tantalum foil lining in a pyrolytically-coated graphite furnace permitted determination of low ppm levels of cerium in most silicate rocks. A preliminary preconcentration by oxalate and hydroxide co-precipitations was used before
Tantalum foil cuffs in peripheral nerve surgery.
E E CLIFFTON
Surgery, 23(3), 507-514 (1948-03-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service