Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF22258278

Nickel

foil, not light tested, 150x150mm, thickness 0.009mm, 99.95%

Synonym(s):

Nickel, NI000190

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$840.00

$840.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$840.00

About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

$840.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 222-582-78

resistivity

6.97 μΩ-cm, 20°C

size × thickness

150 x 150 mm × 0.009 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Kim et al.
Applied physics letters, 98(26), 263106-2631063 (2011-07-30)
Microwave plasma chemical vapor deposition (MPCVD) was employed to synthesize high quality centimeter scale graphene film at low temperatures. Monolayer graphene was obtained by varying the gas mixing ratio of hydrogen and methane to 80:1. Using advantages of MPCVD, the
R H Lowndes et al.
Journal of microscopy, 142(Pt 3), 371-374 (1986-06-01)
A light microscope heating stage suitable for biological temperatures is described. A novel feature of the design is the use of cupro-nickel foil heaters (0.1 mm thick). With a temperature sensor mounted on the slide clip and a temperature controller
Alex Ignatiev et al.
Dalton transactions (Cambridge, England : 2003), (40)(40), 5501-5506 (2008-12-17)
Nanostructured thin film solid oxide fuel cells (SOFC) have been developed for reduced temperature operation, with high power density, and to be self reforming. A thin film electrolyte (1-2 microm thickness), e.g., yttria-stabilized zirconia (YSZ), is deposited on a nickel
L A van Loon et al.
Journal of oral pathology, 17(3), 129-137 (1988-03-01)
An in vivo comparison was made between the contact allergic stomatitis-inducing capacity of nickel, nickel-containing dental alloys and a non-corrosive precious metal. Fifteen patients with a positive allergic skin reaction to nickel were divided into 3 groups (A, B and
H T Tien et al.
Photochemistry and photobiology, 49(5), 527-530 (1989-05-01)
Visible light of the solar spectrum is directly converted to stored chemical energy of hydrogen from artificial sea water in a novel electrochemical photovoltaic cell. The principal element of the cell, modeled after the photosynthetic thylakoid membrane, is a semiconductor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service