Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF21986747

Zirconium

foil, 25x25mm, thickness 0.2mm, annealed, 99.2%

Synonym(s):

Zirconium, ZR000290

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

assay

99.20%

form

foil

manufacturer/tradename

Goodfellow 219-867-47

resistivity

40 μΩ-cm, 20°C

size × thickness

25 x 25 mm × 0.2 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oluwatayo F Ikotun et al.
Future medicinal chemistry, 3(5), 599-621 (2011-04-30)
Positron emission tomography, with its high sensitivity and resolution, is growing rapidly as an imaging technology for the diagnosis of many disease states. The success of this modality is reliant on instrumentation and the development of effective and novel targeted
Abnormal trace metals in man: zirconium.
H A Schroeder et al.
Journal of chronic diseases, 19(5), 573-586 (1966-05-01)
Yugo Hatai
The international journal of esthetic dentistry, 9(2), 206-222 (2014-04-26)
In contemporary dentistry, we have a vast range of materials to choose from, and metal free restorations have become the premier materials for achieving the ultimate in both esthetics and durability. Metal-free restorations are utilized with more conservative preparations to
R Velázquez-Cayón et al.
Medicina oral, patologia oral y cirugia bucal, 17(2), e246-e250 (2011-12-07)
The increase of aesthetic demands, together with the successful outcome of current implants, has renewed interest in the search for new materials with enough mechanical properties and better aesthetic qualities than the materials customarily used in implanto-prosthetic rehabilitation. Among these
Glypican-3-targeted 89Zr PET imaging of hepatocellular carcinoma: where antibody imaging dares to tread.
Bernadette V Marquez et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(5), 708-709 (2014-03-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service