Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF20344407

Nitinol alloy, Ni55Ti45

500mm tube, 0.33mm outside diameter, 0.045mm wall thickness, 0.24mm inside diameter, superelastic

Synonym(s):

NI207050

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23
Product GF20344407 is not currently sold in your country. Contact Technical Service

form

tubes

manufacturer/tradename

Goodfellow 203-444-07

L × thickness

500 mm × 0.045 mm

O.D. × I.D.

0.33 mm × 0.24 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service