GF19192585
Stainless Steel - AISI 316L alloy, FeCr18Ni10Mo3
foil, 90 x 210mm, 0.9mm thickness, annealed, mirror polished on one side
Synonym(s):
FF210375
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352300
NACRES:
NA.23
form
annealed (mirror polished on one side)
foil
manufacturer/tradename
Goodfellow 191-925-85
size × thickness
90 mm × 210 mm × 0.9 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service