Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF18630727

Silver

microfoil, disks, 25mm, thinness 1.0μm, specific density 1098μg/cm2, permanent mylar 3.5μm support, 99.95%

Synonym(s):

Silver, AG004850

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Product GF18630727 is not currently sold in your country. Contact Technical Service

assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 186-307-27

resistivity

1.59 μΩ-cm, 20°C

diam. × thickness

25 mm × 1.0 μm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pakvirun Thuesombat et al.
Ecotoxicology and environmental safety, 104, 302-309 (2014-04-15)
With the advances in nanotechnology, silver nanoparticles (AgNPs) have been applied in many industries, increasing their potential exposure level in the environment, yet their environmental safety remains poorly evaluated. The possible effects of different sized AgNPs (20, 30-60, 70-120 and
Neelam Gogoi et al.
Journal of nanoscience and nanotechnology, 14(6), 4147-4155 (2014-04-18)
In this work storage of silver nanoparticles (Ag NPs) in chitosan gel and its subsequent release for catalytic reduction processes is investigated. The generation of small sized metal nanoparticles which acts as catalyst is prerequisite to progress of a catalytic
Takayoshi Kakuta et al.
Journal of nanoscience and nanotechnology, 14(6), 4090-4096 (2014-04-18)
Three-typed porphyrin derivatives with a different chain-length alkylcarboxylic acid as their peripheral anchor group have been prepared. Anodic photocurrents were observed in a simple system where the porphyrin derivatives were directly anchored on an indium tin oxide (ITO) electrode. Cathodic
Nagaraj Basavegowda et al.
Journal of nanoscience and nanotechnology, 14(6), 4377-4382 (2014-04-18)
The present investigation demonstrates a rapid biogenic approach for the synthesis of gold and silver nanoparticles using biologically active and medicinal important Perilla frutescens leaf extract as a reducing and stabilizing agent under ambient conditions. Gold and silver nanoparticles were
Yanan Ma et al.
Journal of nanoscience and nanotechnology, 14(6), 4245-4250 (2014-04-18)
The properties of the localized surface plasmon resonance (LSPR) and the surface enhanced Raman scattering (SERS) of the core-shell bimetallic nanostructures, that is the monodisperse Au@Ag core-shell nanorods with different thickness of Ag shell, are theoretically and experimental researched. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service