Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF18433740

Tantalum

wire reel, 1m, diameter 0.30mm, annealed, 99.9%

Synonym(s):

Tantalum, TA005135, Ta

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$281.00

$281.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$281.00

About This Item

Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23

$281.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

vapor pressure

<0.01 mmHg ( 537.2 °C)

assay

99.90%

form

wire

autoignition temp.

572 °F

manufacturer/tradename

Goodfellow 184-337-40

resistivity

13.5 μΩ-cm, 20°C

L × diam.

1 m × 0.30 mm

bp

5425 °C (lit.)

mp

2996 °C (lit.)

density

16.69 g/cm3 (lit.)

SMILES string

[Ta]

InChI

1S/Ta

InChI key

GUVRBAGPIYLISA-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
R M Rabinovich
Vestnik rentgenologii i radiologii, (1)(1), 77-79 (1981-01-01)
Clinical use of porous tantalum in complex primary total knee arthroplasty.
Neil P Sheth et al.
American journal of orthopedics (Belle Mead, N.J.), 38(10), 526-530 (2009-12-17)
Metallosis and elevated serum levels of tantalum following failed revision hip arthroplasty--a case report.
George C Babis et al.
Acta orthopaedica, 85(6), 677-680 (2014-08-21)
Giulio Maccauro et al.
Recent patents on biotechnology, 3(3), 157-165 (2009-06-24)
Tantalum is obtained from the minerals colombite, tantalite and euxenite. It is greyish silver, heavy and very hard. Tantalum does not react with body fluids and is used to make surgical equipment. Tantalum also does not irritate the body and
Juncai Deng et al.
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi, 28(2), 419-422 (2011-05-25)
This paper describes the biological characteristics of the metal tantalum (Ts) and the application status of tantalum in artificial joints, bone necrosis, spine, defects of bone and other aspects of bone. The early clinical application results of tantalum in bone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service