GF16165919
Tantalum
wire, straight, 500mm, diameter 0.5mm, annealed, 99.9%
Synonym(s):
Tantalum, TA005350, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.01 mmHg ( 537.2 °C)
assay
99.90%
form
wire
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 161-659-19
resistivity
13.5 μΩ-cm, 20°C
L × diam.
500 mm × 0.5 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[Tantalum dust as a contrast substance for laryngography and bronchography (a review of the literature)].
R M Rabinovich
Vestnik rentgenologii i radiologii, (1)(1), 77-79 (1981-01-01)
Xiao-kang Tang et al.
Zhongguo gu shang = China journal of orthopaedics and traumatology, 26(7), 617-620 (2013-10-19)
Incorrect treatment for femur head necrosis can cause collapse of femoral head and tresult in severe harm for the patients (especially for the patient with middle-aged and young). The structure and mechanics characteristics of tantalum rod is similar to bone
Ulrich Dorn et al.
The Journal of arthroplasty, 29(4), 831-835 (2013-10-09)
This study compared the corrosion behavior of tantalum-coated cobalt-chromium modular necks with that of titanium alloy modular necks at their junction to titanium-alloy femoral stem. Tests were performed in a dry assembly and two wet assemblies, one contaminated with calf
Clinical use of porous tantalum in complex primary total knee arthroplasty.
Neil P Sheth et al.
American journal of orthopedics (Belle Mead, N.J.), 38(10), 526-530 (2009-12-17)
Robert Cohen
American journal of orthopedics (Belle Mead, N.J.), 31(4), 216-217 (2002-05-15)
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service