Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF13791708

Iron

rod, 50mm, diameter 8.0mm, as drawn, 99.99+%

Synonym(s):

Iron, FE007948

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$689.00

$689.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$689.00

About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
12141721
PubChem Substance ID:
NACRES:
NA.23

$689.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

99.99%

form

rod

manufacturer/tradename

Goodfellow 137-917-08

resistivity

9.71 μΩ-cm

L × diam.

50 mm × 8.0 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Wilhelm et al.
Clinical nephrology, 81(4), 251-258 (2014-03-25)
Hyperphosphatemia in advanced chronic kidney disease (CKD) necessitates the use of phosphate binders. This in vitro study assessed phosphate binding and Fe release properties of the novel iron-based phosphate binder PA21. Phosphate adsorption and Fe release were assessed under conditions
S J Ford et al.
Annals of the Royal College of Surgeons of England, 96(4), 275-278 (2014-05-02)
The incidence of oesophageal adenocarcinoma (OAC) is rising dramatically and overall survival remains extremely poor. Iron has been shown to potentiate tumourigenesis in OAC, and iron chelation therapy demonstrates promise in vivo as an adjunct to neoadjuvant and palliative chemotherapy.
Yukio Miyamoto et al.
AJR. American journal of roentgenology, 202(4), W400-W407 (2014-03-26)
The objective of our study was to compare the efficacy of contrast-enhanced ultrasound (CEUS) using the ultrasound contrast agent Sonazoid (perflubutane) with unenhanced ultrasound and supplementary contrast-enhanced MRI in the differential diagnosis (benign vs malignant) of focal breast lesions. The
Lucia Banci et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(17), 6203-6208 (2014-04-16)
Monothiol glutaredoxins play a crucial role in iron-sulfur (Fe/S) protein biogenesis. Essentially all of them can coordinate a [2Fe-2S] cluster and have been proposed to mediate the transfer of [2Fe-2S] clusters from scaffold proteins to target apo proteins, possibly by
Syllos S da Silva et al.
Environmental technology, 35(9-12), 1556-1564 (2014-04-08)
The photo-Fenton process was applied to degrade non-ionic surfactants with different numbers of ethoxy groups, seven (E7), ten (E10) and twenty-three (E23). The effects of H2O2 concentration, Fe(II) concentration and number of ethoxy groups on the mineralization of surfactants were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service