Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF12090394

Zinc

foil, 100x100mm, thickness 0.125mm, as rolled, 99.95+%

Synonym(s):

Zinc, ZN000270

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 120-903-94

resistivity

5.8 μΩ-cm, 20°C

size × thickness

100 x 100 mm × 0.125 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Haimin Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(17), 5165-5169 (2012-03-21)
Tandra Ghoshal et al.
Journal of nanoscience and nanotechnology, 9(9), 5586-5591 (2009-11-26)
Nanowire arrays of zinc oxide were synthesized on zinc foil by a simple thermal evaporation process. Morphologies and sizes of the synthesized nanostructures were varied by varying the reaction time and the surface roughness of the substrate. Self-catalytic liquid-solid mechanism
S Rondot et al.
Science (New York, N.Y.), 263(5154), 1739-1741 (1994-03-25)
An x-ray projection microscope equipped with a charge-coupled device camera allows direct observation of zinc (Zn(2+)) ions diffusing in aqueous hydrochloric acid solution during the corrosion of zinc foil and pellets. Time series of microradiographic images with a lateral resolution
Chenglin Yan et al.
The journal of physical chemistry. B, 110(51), 25850-25855 (2006-12-22)
Vertically aligned ZnO nanorods with uniform diameter and length have been synthesized on a zinc foil substrate with ammonium persulfate as oxidant via a facile, larger scale production and inexpensively synthesized method without any templates or additives. SEM and XRD
Xufeng Wu et al.
Chemical communications (Cambridge, England), (15)(15), 1655-1657 (2006-04-04)
Highly oriented ZnO nanoneedle/nanorods arrays have been fabricated by direct oxidation of zinc foil in alkaline zincate ion solution at near room temperature (20 degrees C for nanoneedles, 30 degrees C for nanorods).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service