Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF10620841

Titanium

tube, outside diameter 7.0 mm, length 200 mm, wall thickness 1.0 mm

Synonym(s):

TI007370

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$484.00
5 EA
$1,960.00

$484.00


Estimated to ship onMay 19, 2025FromMILWAUKEE


Request a Bulk Order

Select a Size

Change View
1 EA
$484.00
5 EA
$1,960.00

About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
MDL number:
UNSPSC Code:
12141746
PubChem Substance ID:
NACRES:
NA.23

$484.00


Estimated to ship onMay 19, 2025FromMILWAUKEE


Request a Bulk Order

form

tubes

autoignition temp.

860 °F

manufacturer/tradename

Goodfellow 106-208-41

resistivity

42.0 μΩ-cm, 20°C

L

200 mm

O.D.

7.0 mm

wall thickness

1.0 mm

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

SMILES string

[Ti]

InChI

1S/Ti

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service