Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF08507257

Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3

sphere, 5mm diameter, 2.50μm sphericity, precision sphere, grade 100, 50pcs

Synonym(s):

FE246850

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
NACRES:
NA.23

form

spheres (grade 100)

quality

precision sphere

packaging

1 ea of 50 pieces

manufacturer/tradename

Goodfellow 085-072-57

diam.

5 mm

sphericity

2.50 μm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service