Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF06938657

Platinum

wire reel, 10m, diameter 0.063mm, hard, 99.9%

Synonym(s):

Platinum, PT005112

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$751.00

$751.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$751.00

About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

$751.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

assay

99.9%

form

wire

manufacturer/tradename

Goodfellow 069-386-57

resistivity

10.6 μΩ-cm, 20°C

L × diam.

10 m × 0.063 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio González
Future oncology (London, England), 9(12 Suppl), 29-35 (2013-11-15)
Ovarian carcinoma is still one of the most common causes of death from cancer in the western world. Despite high sensitivity to chemotherapy, the majority of patients will relapse within 3 years. In this article the focus is centered on
Nicoletta Colombo
Future oncology (London, England), 9(12 Suppl), 19-23 (2013-11-15)
Ovarian cancer is the leading cause of gynecological cancer deaths worldwide. Despite primary treatment with platinum-containing regimens, the majority of women will experience recurrent disease and subsequent death. Recurrent ovarian cancer remains a challenge for successful management, and the choice
Thorsten Langer et al.
Trends in pharmacological sciences, 34(8), 458-469 (2013-06-19)
Childhood cancer survival rates are now nearly 80% in more developed European countries because of improved therapies and better supportive care. Platinum chemotherapy drugs, such as cisplatin and carboplatin, are the cornerstone of many effective therapeutic protocols for childhood cancer.
Himashinie V K Diyabalanage et al.
Cancer letters, 329(1), 1-8 (2012-10-04)
One of the most promising strategies to increase the efficacy of standard chemotherapy drugs is by combining them with low doses of histone deacetylases inhibitors (HDACis). Regarded as chemosensitizers, the addition of well-tolerated doses of HDACis to platinum-based chemotherapeutics has
Hai-Bo Wei et al.
Chinese medical journal, 125(16), 2902-2907 (2012-08-31)
Platinum-based regimens are used as standard first-line chemotherapy in non-small cell lung cancer (NSCLC) patients. To study if pharmacogenetic approach may allow a tailored selection of platinum chemotherapy for advanced NSCLC, we performed a meta-analysis to compare chemosensitivity to platinum

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service