GF03140862
Stainless steel - AISI 304
foil, Fe/Cr18%/Ni 10%, thickness 1.0 mm, size 25 × 25 mm, temper annealed
Synonym(s):
FE220360
About This Item
form
foil
manufacturer/tradename
Goodfellow 031-408-62
size
25 × 25 mm
thickness
1.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
signalword
Danger
hcodes
pcodes
Hazard Classifications
Carc. 2 - Skin Sens. 1 - STOT RE 1
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service