Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF02814702

Beryllium

mesh, 5x5mm, nominal aperture 0.13mm, thickness 0.025mm, wire diameter 0.1mm, 100 wires/inch, open area 50%, electro-formed mesh

Synonym(s):

Beryllium, BE008710, Glucinium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Be
CAS Number:
Molecular Weight:
9.01
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

mesh

autoignition temp.

1198 °F

manufacturer/tradename

Goodfellow 028-147-02

resistivity

4.46 μΩ-cm, 20°C

L × W

5 mm × 5 mm

thickness × diam.

0.025 mm × 0.1 mm

bp

2970 °C (lit.)

mp

1278 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

SMILES string

[Be]

InChI

1S/Be

InChI key

ATBAMAFKBVZNFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce C Wainman et al.
Chemosphere, 149, 190-201 (2016-02-09)
Persistent organohalogens (POHs) and metals have been linked to alterations in menstrual cycle function and fertility in humans. The Cree First Nations people living near James Bay in Ontario and Quebec, Canada, have elevated levels of POHs, mercury and lead
Erin Shay et al.
Critical reviews in toxicology, 43(3), 220-243 (2013-03-01)
The exposure-response patterns with beryllium sensitization (BeS), chronic beryllium disease (CBD) and lung cancer are influenced by a number of biological and physicochemical factors. Recent studies have suggested dermal exposure as a pathway for BeS. In light of the current
Kathleen Kreiss et al.
Annual review of public health, 28, 259-277 (2006-11-11)
Beryllium exposure can cause a granulomatous lung disease in workers who develop a lymphocyte-mediated sensitization to the metal. Workers in diverse industries are at risk because beryllium's properties are critical to nuclear, aerospace, telecommunications, electronic, metal alloy, biomedical, and semiconductor
K D Rosenman et al.
Occupational and environmental medicine, 68(7), 487-493 (2010-12-28)
Chronic beryllium disease (CBD) is a hypersensitivity granulomatous pulmonary disease caused by exposure to the metal beryllium (Be²⁺). Our objective was to extend current knowledge of the genetics of beryllium disease by examining all HLA-DPB1 and HLA-DPR1 gene polymorphisms and
Richard T Sawyer et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 24(1), 1-17 (2010-10-29)
During the last decade, there have been concerted efforts to reduce beryllium (Be) exposure in the workplace and thereby reduce potential cases of this occupational lung disorder. Despite these efforts, it is estimated that there are at least one million

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service