GF02784911
Stainless steel - 15-7ph
foil, Fe/Cr 15%/Ni 7%/Mo 2.25%, thickness 0.125 mm, length 2 m, temper annealed
Synonym(s):
FE290320
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.23
Recommended Products
form
foil
manufacturer/tradename
Goodfellow 027-849-11
L
2 m
thickness
0.125 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service