Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF02621225

Copper-beryllium alloy, Cu98Be2

foil, 90 x 100mm, 0.0065mm thickness, as rolled, light tight (lt)

Synonym(s):

CU070230

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
31152304
NACRES:
NA.23
Pricing and availability is not currently available.

description

light tight (lt)

form

foil (as rolled)

manufacturer/tradename

Goodfellow 026-212-25

size × thickness

90 mm × 100 mm × 0.0065 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service