GF00662363
Boron
powder, mean particle size (fsss) 1 micron, weight 200 g, purity 92.1%
Synonym(s):
B 006020
About This Item
assay
92.1%
form
powder
manufacturer/tradename
Goodfellow 006-623-63
resistivity
1.5E12 μΩ-cm, 20 °C
weight
200 g
mean particle size
1 μm (FSSR)
density
2.34 g/mL at 25 °C (lit.)
SMILES string
[B]
InChI
1S/B
InChI key
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service