Key Documents
GE80-6467-22
GE Ettan DALT
twelve gel caster system
About This Item
UNSPSC Code:
41100000
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 80-6467-22
availability
not available in Japan
General description
Hoefer DALT System Function and Description
In 2-D protein electrophoresis, proteins are separated first according to isoelectric point by isoelectric focusing, most reliably on immobilized pH gradient (IPG) strips. The second dimension electrophoresis separates the proteins on the basis of their molecular mass on a slab gel containing the denaturing detergent, sodium dodecyl sulfate (SDS).
The Hoefer DALT System is designed to simplify the handling of multiple second dimension gels and improve the reproducibility of the second dimension separation.
DALT System Components
  • a multiple vertical slab electrophoresis tank
  • a multiple gel caster
  • a gradient maker with peristaltic pump
  • gel cassettes
  • a blotting kit
Legal Information
Ettan is a trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)