Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F6062

Sigma-Aldrich

Fmoc-(S)-2-(2-propenyl)Ala-OH

Synonym(s):

(S)-N-Fmoc-α-2-n-propenylalanine, Fmoc-(S)-2-(2-propenyl)alanine, Fmoc-(S)-2-amino-2-methyl-4-pentenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥97%

Quality Level

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

O=C(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)N[C@](CC=C)(C)C(O)=O

InChI

1S/C21H21NO4/c1-3-12-21(2,19(23)24)22-20(25)26-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h3-11,18H,1,12-13H2,2H3,(H,22,25)(H,23,24)/t21-/m0/s1

InChI key

FNCSRFHDUZYOCR-NRFANRHFSA-N

Application

Olefinic alpha-methyl amino acid for peptide stapling. Upon incorporation of this amino acid into a peptide, along with another of the same or derivative with a different length of the olefinic side chain, the two can be ′stapled′ via a ring closing metathesis reaction with Grubb′s catalyst (product # 579726). The resulting stapled peptide macrocycle has been shown to stabilize the alpha-helical structure of peptides, which can lead to favorable biological characteristics such as increased proteolytic stability and cellular uptake.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Woo Kim et al.
Nature protocols, 6(6), 761-771 (2011-06-04)
This protocol provides a detailed procedure for the preparation of stapled α-helical peptides, which have proven their potential as useful molecular probes and as next-generation therapeutics. Two crucial features of this protocol are (i) the construction of peptide substrates containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service