F5562
Fmoc-(S)-2-(2-azidoethane)Ala-OH
≥97%
Synonym(s):
(S)-2-{[(9H-fluoren-9-yl)methoxy]carbonylamino}-4-azido-2-methylbutanoic acid, Fmoc-(S)-4-azido-2-methylbutanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H20N4O4
CAS Number:
Molecular Weight:
380.40
UNSPSC Code:
12352200
NACRES:
NA.22
assay
≥97%
reaction suitability
reaction type: click chemistry
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
−20°C
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service