Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E9641

Sigma-Aldrich

Ethyl acetoacetate

99%

Synonym(s):

Acetoacetic ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC2H5
CAS Number:
Molecular Weight:
130.14
Beilstein/REAXYS Number:
385838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

4.48 (vs air)

vapor pressure

1 mmHg ( 28.5 °C)

assay

99%

autoignition temp.

580 °F

expl. lim.

9.5 %

bp

181 °C (lit.)

mp

−43 °C (lit.)

density

1.029 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)CC(C)=O

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

XYIBRDXRRQCHLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gui-Rong Qu et al.
Organic letters, 11(8), 1745-1748 (2009-03-20)
A novel approach to the synthesis of purines bearing functionalized carbon substituents or methyl in position 6 was developed. Under different reaction conditions, 6-halopurine derivatives could react with ethyl acetoacetate efficiently to yield 2-(purin-6-yl)acetoacetic acid ethyl esters, (purin-6-yl)acetates and 6-methylpurines
Arne T Dickschat et al.
Chemical communications (Cambridge, England), 49(22), 2195-2197 (2013-02-09)
Bifunctional mesoporous silica nanoparticles (MSNs) bearing Pd-complexes and additional basic sites were prepared and tested as cooperative active catalysts in the Tsuji-Trost allylation of ethyl acetoacetate. Functionalization of the MSNs was realized by postmodification using click-chemistry. The selectivity of mono
Hassan Valizadeh et al.
Molecular diversity, 15(1), 233-237 (2010-07-24)
In this study, a three-component one-pot synthesis of select 5-amino-6-cyano-3-hydroxybenzo[c]coumarin compounds derived from salicylaldehydes, malononitrile, and ethyl acetoacetate is reported. The reaction is conducted on grinding over MgO at room temperature resulting in good yields.
Debasish Bandyopadhyay et al.
Molecules (Basel, Switzerland), 17(3), 2643-2662 (2012-03-07)
The classical Hantzsch reaction is one of the simplest and most economical methods for the synthesis of biologically important and pharmacologically useful 1,4-dihydropyridine derivatives. Bismuth nitrate pentahydrate under microwave irradiation is proven to act as a very efficient catalyst for
Junguo Xin et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(10), 3177-3181 (2008-02-05)
An enantioselective Biginelli reaction that proceeds by a dual-activation route has been developed by using a combined catalyst of a readily available trans-4-hydroxyproline-derived secondary amine and a Brønsted acid. Aromatic, heteroaromatic, and fused-ring aldehydes were found to be suitable substrates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service