Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E28404

Sigma-Aldrich

1-Pentyn-3-ol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(OH)C≡CH
CAS Number:
Molecular Weight:
84.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

1 (vs air)

assay

98%

refractive index

n20/D 1.434 (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CCC(O)C#C

InChI

1S/C5H8O/c1-3-5(6)4-2/h1,5-6H,4H2,2H3

InChI key

LBSKEFWQPNVWTP-UHFFFAOYSA-N

pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

84.2 °F - closed cup

flash_point_c

29 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Killary et al.
Experimental cell research, 154(2), 442-453 (1984-10-01)
A selective system toxic towards mammalian cells expressing the liver-specific isozyme of alcohol dehydrogenase (L-ADH) has been developed. A number of alpha-unsaturated primary and secondary alcohols were assayed for their ability to serve as substrates for rat liver ADH and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service