Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E22805

Sigma-Aldrich

Ethyl 2,3-dibromopropionate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrCHBrCOOC2H5
CAS Number:
Molecular Weight:
259.92
Beilstein/REAXYS Number:
636046
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.5 (lit.)

bp

211-214 °C/746 mmHg (lit.)

density

1.788 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(Br)CBr

InChI

1S/C5H8Br2O2/c1-2-9-5(8)4(7)3-6/h4H,2-3H2,1H3

Inchi Key

OENICUBCLXKLJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service