Based on my research, I couldn't find any specific instructions for using solution C4361 for cell culture purposes. However, I did come across a product information sheet that describes the use of C4361 for a gelatinase assay, with no mention of its use for cell culture.
DWKW216936
Wheaton wide-mouth bottle
without cap, wide-mouth amber soda-lime glass bottle, capacity (500 mL)
Synonym(s):
W216936
Select a Size
$53.00
Available to ship onApril 29, 2025Details
Select a Size
About This Item
$53.00
Available to ship onApril 29, 2025Details
Recommended Products
material
wide-mouth amber soda-lime glass bottle
sterility
non-sterile
feature
with cap
without cap
packaging
case of 24 ea
manufacturer/tradename
WHEATON® W216936
bottle diam. × H × W
(79 mm) (133 mm) (319 mm)
capacity
(500 mL)
fitting
63-400 neck
63-400
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Available in larger bottle sizes
- Wide mouth / neck
- Clear, Type III soda-lime glass
- Available with caps attached or without caps
- HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification
- Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What are the dimensions of the bottle for item DWKW216936, the Wheaton wide-mouth bottle?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service