Key Documents
DWK247716906
DURAN® desiccator base with plain flange and NS-tubulature
Synonym(s):
247716906
About This Item
UNSPSC Code:
41104510
NACRES:
NA.54
Product DWK247716906 is not currently sold in your country. Contact Technical Service
material
borosilicate glass 3.3
joint
joint ST/NS hose barb24/29
packaging
case of 1 ea
manufacturer/tradename
DURAN® 247716906
H
283 mm
flange I.D.
332 mm
General description
Standard ground outlet (24/29), type NOVUS, suitable for all types of lids
Features and Benefits
  • Complies with DIN ISO 13130
  • Manufactured from borosilicate glass 3.3
  • Tight closure due to standard ground joint
  • NOVUS type with standard ground joint (NS 24/29) junction tube in the lid
  • With flat flange
  • Suitable for all types of lids
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)