Key Documents
DWK218202404
DURAN® wide mouth laboratory bottle
without cap, square glass bottle, bottle capacity (100 mL), non-sterile
Synonym(s):
218202404
About This Item
UNSPSC Code:
41121800
Product DWK218202404 is not currently sold in your country. Contact Technical Service
material
square glass bottle
sterility
non-sterile
feature
without cap
packaging
case of 10 ea
manufacturer/tradename
DURAN® 218202404
bottle capacity
(100 mL)
fitting
GL 80 cap
GL 80 neck
General description
Easy access to the bottle contents due to the wide diameter of the bottle neck. Optimum filling and pouring and particularly easy cleaning. Spoons, spatulas and tweezers can be manipulated with ease. Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base area. Easy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic. Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)