Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D24005

Sigma-Aldrich

2,3-Diaminopropionic acid monohydrobromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8N2O2 · HBr
CAS Number:
Molecular Weight:
185.02
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98%

SMILES string

Br.NCC(N)C(O)=O

InChI

1S/C3H8N2O2.BrH/c4-1-2(5)3(6)7;/h2H,1,4-5H2,(H,6,7);1H

InChI key

IZNPALADKWAZMW-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service