Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

D204838

Sigma-Aldrich

2-Diphenylacetyl-1,3-indandione-1-hydrazone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H18N2O2
CAS Number:
Molecular Weight:
354.40
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

assay

98%

form

powder, crystals or chunks

mp

241-243 °C (lit.)

solubility

acetone: 2 mg/mL, clear, yellow to brown

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N\N=C1\C(C(=O)C(c2ccccc2)c3ccccc3)C(=O)c4ccccc14

InChI

1S/C23H18N2O2/c24-25-21-17-13-7-8-14-18(17)22(26)20(21)23(27)19(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,19-20H,24H2/b25-21+

InChI key

QBGMRRUHCFHZLC-NJNXFGOHSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Limitations of 2-diphenylacetyl-1,3-indandione-1-hydrazone as a precolumn fluorescence derivatization reagent.
M A Heindorf et al.
Journal of chromatography, 464(1), 186-194 (1989-02-17)
[Quantitative determination of oxoline].
Z B Moriak et al.
Farmatsevtychnyi zhurnal, (1)(1), 70-70 (1980-01-01)
J M Rideout et al.
Clinica chimica acta; international journal of clinical chemistry, 161(1), 29-35 (1986-11-30)
A simple sensitive method for the assay of blood acetaldehyde is described. Samples are collected into an organic solution of 2-diphenylacetyl-1,3-indandione-1-hydrazone. Proteins are precipitated immediately and carbonyl compounds react to form fluorescent azine derivatives. Methanol in the reagent mixture prevents
Effects of ethanol and glutamate agonist infusion on the outflow of sulphoacetaldehyde: microdialysis studies.
S Capodarca et al.
Advances in experimental medicine and biology, 483, 297-304 (2002-01-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service