Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D180300

Sigma-Aldrich

cis-2,6-Dimethylpiperidine

98%

Synonym(s):

2,6-Lupetidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N
CAS Number:
Molecular Weight:
113.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

98%

form

liquid

refractive index

n20/D 1.4394 (lit.)

bp

127-128 °C/768 mmHg (lit.)

density

0.84 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1CCC[C@@H](C)N1

InChI

1S/C7H15N/c1-6-4-3-5-7(2)8-6/h6-8H,3-5H2,1-2H3/t6-,7+

InChI key

SDGKUVSVPIIUCF-KNVOCYPGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for elimination reactions

Reactant for corrosion inhibitors for iron in HCl

Catalyst for:
Preparation of aliphatic amines
Ketene forming eliminations
Stereoselective synthesis of triflates

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Z Fan et al.
Nanoscale, 11(22), 10774-10781 (2019-05-28)
Colloidal quantum dots have garnered significant interest in optoelectronics, particularly in quantum dot solar cells (QDSCs). Here we report QDSCs fabricated using a ligand that is modified, following film formation, such that it becomes an efficient hole transport layer. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service