Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D1534

Sigma-Aldrich

Diethylenetriaminepentaacetic acid α,ω-bis(biocytinamide)

≥95% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H75N11O16S2
CAS Number:
Molecular Weight:
1102.28
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥95% (TLC)

form

solid

storage temp.

2-8°C

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(=O)NC(CCCCNC(=O)CCCCC1SCC2NC(=O)NC12)C(O)=O)CCN(CC(O)=O)CC(=O)NC(CCCCNC(=O)CCCCC3SCC4NC(=O)NC34)C(O)=O

InChI

1S/C46H75N11O16S2/c58-34(13-3-1-11-32-41-30(26-74-32)51-45(72)53-41)47-15-7-5-9-28(43(68)69)49-36(60)21-56(24-39(64)65)19-17-55(23-38(62)63)18-20-57(25-40(66)67)22-37(61)50-29(44(70)71)10-6-8-16-48-35(59)14-4-2-12-33-42-31(27-75-33)52-46(73)54-42/h28-33,41-42H,1-27H2,(H,47,58)(H,48,59)(H,49,60)(H,50,61)(H,62,63)(H,64,65)(H,66,67)(H,68,69)(H,70,71)(H2,51,53,72)(H2,52,54,73)

InChI key

LKGHKLSIBPOASY-UHFFFAOYSA-N

Application

A metal chelating agent which may be site-directed by using the biotin-avidin binding system.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Hnatowich et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 28(8), 1294-1302 (1987-08-01)
The attractive properties of avidin (streptavidin) and biotin, in particular their strong affinities (Kd = 10(-15)M), may be used to advantage in imaging applications. These molecules have been used in this preliminary investigation to improve the targeting of 111In in
D J Hnatowich et al.
Science (New York, N.Y.), 220(4597), 613-615 (1983-05-06)
A simple and efficient method of covalently coupling the strong chelator diethylenetriaminepentaacetic acid to proteins was developed for radiolabeling immunoglobulin G antibodies. After being coupled and labeled with indium-111, a monoclonal antibody to carcinoembryonic antigen retained its ability to bind

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service