Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D152951

Sigma-Aldrich

Dimethyl chlorothiophosphate

97%

Synonym(s):

Dimethyl phosphorochloridothionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(S)Cl
CAS Number:
Molecular Weight:
160.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

0.67 psi ( 20 °C)

assay

97%

form

liquid

refractive index

n20/D 1.482 (lit.)

bp

66-67 °C/16 mmHg (lit.)

density

1.322 g/mL at 25 °C (lit.)

SMILES string

COP(Cl)(=S)OC

InChI

1S/C2H6ClO2PS/c1-4-6(3,7)5-2/h1-2H3

InChI key

XFBJRFNXPUCPKU-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

203.0 °F - open cup

flash_point_c

95 °C - open cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolaos I Rousis et al.
Water research, 121, 270-279 (2017-05-30)
Human biomonitoring, i.e. the determination of chemicals and/or their metabolites in human specimens, is the most common and potent tool for assessing human exposure to pesticides, but it suffers from limitations such as high costs and biases in sampling. Wastewater-based
Mateusz Daśko et al.
Drug development research, 80(6), 857-866 (2019-07-14)
In the present work, we described convenient methods for the synthesis of N-thiophosphorylated 3-(4-aminophenyl)-coumarin-7-O-sulfamates as steroid sulfatase (STS) inhibitors. To design the structures of the potential STS inhibitors, molecular modeling techniques were used. A computational docking method was used to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service