Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CVT00092

Sigma-Aldrich

3-(Bromomethyl)-5-(3-(trifluoromethyl)phenyl)-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8BrF3N2
Molecular Weight:
305.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

SMILES string

BrCC1=NNC(C2=CC=CC(C(F)(F)F)=C2)=C1

InChI

1S/C11H8BrF3N2/c12-6-9-5-10(17-16-9)7-2-1-3-8(4-7)11(13,14)15/h1-5H,6H2,(H,16,17)

InChI key

RSUWRQUDCNGPPG-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service