CLS7103C20
Corning® volumetric pipette, TC/TD, SER/CRT, Corning® 7103C
Synonym(s):
pipet
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121510
material
glass
feature
class A
graduation
manufacturer/tradename
Corning 7103C20
availability
available only in South Korea
volume measuring range
20 mL, accuracy: 0.03 mL mL
capacity
20 mL
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service