Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS7103C15

Corning® volumetric pipette, TC/TD, SER/CRT, Corning® 7103C

Synonym(s):

pipet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121510
Pricing and availability is not currently available.

material

glass

feature

class A
graduation

manufacturer/tradename

Corning 7103C15

availability

available only in South Korea

volume measuring range

15 mL, accuracy: 0.03 mL

capacity

15 mL

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service