Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS6982516X

PYREXPLUS® culture tube w/ screw-cap and protective coating

capacity 15.0 mL

Synonym(s):

culture tubes, test tubes, test tubes glass, Corning® 69825

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

cap (Rubber liner)
round bottom tube

packaging

case of 144

manufacturer/tradename

Corning 69825-16X

O.D. × L

16 mm × 125 mm

capacity

15.0 mL

fitting

15-415 thread

Looking for similar products? Visit Product Comparison Guide

General description

Tube have a protective polymer coating to prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. Deep-form phenolic caps included.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service