Key Documents
CLS3952530C
Pyrex® gas dispersion fritted disc
porosity 40-60 μm
Synonym(s):
Corning® 39525
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
manufacturer/tradename
Corning 39525-30C
L
250 mm
disc diam.
30 mm
pore size
40-60 μm porosity
General description
These tubes, with a fritted disc at an angle of 90°, are used to disperse or scrub gas in liquids.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)