Key Documents
CLS3900
Pyrex® extraction apparatus, arsine generator
complete
Synonym(s):
Corning® 3900
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Product CLS3900 is not currently sold in your country. Contact Technical Service
manufacturer/tradename
Corning 3900
General description
Designed for arsenic determination by the silver diethyldihiocarbramate photometric method. Meets A.P.H.A. Standard 104A, ACS/USP and EPA specifications. Fabricated from Pyrex borosilicate glass. Joint on 125mL flask is 24/40 and ball and socket joint between absorber and scrubber is 12/2. Complete unit includes clamp.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)