Key Documents
CLS353146
Falcon® Petri Dish
diam. × H 90 mm × 15 mm, sterile, polystyrene, with 3 vents, sleeve of 20 × 25 pieces
Synonym(s):
plastic cell culture dish
About This Item
UNSPSC Code:
41122101
Pricing and availability is not currently available.
material
polystyrene
sterility
γ-irradiated (SAL 10-6)
sterile
feature
with 3 vents
packaging
sleeve of 20 × 25 pieces
manufacturer/tradename
Corning
availability
available only in (For sale in India only)
diam. × H
90 mm × 15 mm
Features and Benefits
  • Manufactured under cGMP with optically clear Virgin PS
  • Sterilized to achieve sterility assurance level of 10-6.Sterlity achieved through gamma irradiation
  • Features three vents to provide consistent gas exchange and enhance the stability of stacked dishes
Legal Information
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)