Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CF0034

Sigma-Aldrich

4-Bromophenoxytetrafluoroethanesulphonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H4BrF5O3S
CAS Number:
MDL number:
UNSPSC Code:
12352101

form

liquid

reaction suitability

reaction type: C-C Bond Formation

InChI

1S/C8H4BrF5O3S/c9-5-1-3-6(4-2-5)17-7(10,11)8(12,13)18(14,15)16/h1-4H

InChI key

SPXWBVPRFLQEGR-UHFFFAOYSA-N

Application

4-Bromophenoxytetrafluoroethanesulfonyl fluoride serves as a moderately reactive 4-bromophenoxytetrafluoroethanesulfonylation reagent. Fluoroalkylsulfonylation of the amine nitrogen greatly lowers the pKa value of the N-H and can be used to modulate the behaviour of the drug candidate or build additional molecular complexity around the highly acidic fluoroalkylsulfonamide nitrogen.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service