Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CF0008

Sigma-Aldrich

Alcohol Togni-(PyrazolylCF2CF2)-reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C14H13F4IN2O
CAS Number:
MDL number:
UNSPSC Code:
12352005

form

liquid

reaction suitability

reaction type: C-C Bond Formation

storage temp.

−20°C

Application

Alcohol Togni-(Pyrazolyl-CF2CF2)-reagent is an electrophilic hypervalent iodine-fluoroalkyl reagent introducing the pyrazolyltetrafluoroethyl fragment.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service